siRNA Details
Gene Symbol: DONSON | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017613.3 | NP_060083.1 | 9 | 1529 | ||
GenBank mRNA | AF232673.1 | - | 8 | 1150 | ||
AF232674.1 | - | 7 | 971 | |||
AF232675.1 | - | 4 | 585 | |||
AK001274.1 | - | 7 | 929 | |||
AK074964.1 | - | 9 | 1478 | |||
BC048266.1 | - | 9 | 1479 | 4393399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
