siRNA Details
Gene Symbol: RET | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020630.4 | NP_065681.1 | 1491 | |||
NM_020975.4 | NP_066124.1 | 1491 | ||||
GenBank mRNA | AK291807.1 | - | 1361 | |||
BC004257.1 | - | 1481 | 3160389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
