siRNA Details
Gene Symbol: LOC107987373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724896.2 | XP_006724959.1 | 4 | 351 | ||
XM_017030111.1 | XP_016885600.1 | 4 | 351 |
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021134.3 | NP_066957.3 | 4 | 351 | ||
XM_006718271.2 | XP_006718334.1 | 4 | 351 | |||
XM_011520273.1 | XP_011518575.1 | 4 | 351 | |||
XM_011520275.2 | XP_011518577.1 | 4 | 351 | |||
XM_011520276.2 | XP_011518578.1 | 4 | 351 | |||
GenBank mRNA | AK313805.1 | - | 4 | 291 | ||
BC027710.2 | - | 4 | 266 | 4298000 | ||
U26596.1 | - | 4 | 314 | |||
Z49254.1 | - | 4 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
