siRNA Details
Gene Symbol: ZNF564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144976.3 | NP_659413.1 | 4 | 1794 | ||
GenBank mRNA | AK092760.1 | - | 4 | 1755 | ||
AL834146.1 | - | 4 | 5054 | |||
BC028367.1 | - | 4 | 1731 | |||
BC065023.1 | - | 4 | 1704 | 6044821 | ||
BC136728.1 | - | 4 | 1630 | 9020718 | ||
BC136729.1 | - | 4 | 1630 | 9020719 | ||
BX647959.1 | - | 4 | 1766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
