siRNA Details
Gene Symbol: ADAMTS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197941.3 | NP_922932.2 | 2, 3 | 964 | ||
NR_135689.1 | 2, 3 | 964 | ||||
XM_011543113.2 | XP_011541415.1 | 2, 3 | 979 | |||
XM_011543114.2 | XP_011541416.1 | 2, 3 | 558 | |||
XM_011543115.2 | XP_011541417.1 | 2, 3 | 1017 | |||
XM_011543116.2 | XP_011541418.1 | 2, 3 | 1014 | |||
XM_011543119.2 | XP_011541421.1 | 2, 3 | 1008 | |||
XM_011543120.2 | XP_011541422.1 | 2, 3 | 1005 | |||
XM_011543122.2 | XP_011541424.1 | 2, 3 | 1005 | |||
XM_011543126.2 | XP_011541428.1 | 2, 3 | 1002 | |||
XM_011543127.2 | XP_011541429.1 | 2, 3 | 1002 | |||
GenBank mRNA | AB209685.1 | - | 2, 3 | 461 | ||
AF140674.1 | - | 1, 2 | 128 | |||
AY692424.1 | - | 2, 3 | 922 | |||
AY692425.1 | - | 1, 2 | 126 | |||
AY692426.1 | - | 1, 2 | 125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
