siRNA Details
Gene Symbol: MXRA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008528.2 | NP_001008528.1 | 2, 3 | 472 | ||
NM_001008529.2 | NP_001008529.1 | 2, 3 | 472 | |||
NM_198530.3 | NP_940932.2 | 2, 3 | 472 | |||
NR_130926.1 | 2, 3 | 171 | ||||
NR_130927.1 | 2, 3 | 289 | ||||
NR_130928.1 | 2, 3 | 289 | ||||
XM_005257382.2 | XP_005257439.1 | 1, 2 | 558 | |||
XM_005257383.2 | XP_005257440.1 | 1, 2 | 558 | |||
XM_005257384.2 | XP_005257441.1 | 1, 2 | 558 | |||
XM_011524830.1 | XP_011523132.1 | 2, 3 | 358 | |||
GenBank mRNA | AK128460.1 | - | 1, 2 | 2115 | ||
AW131567.1 | - | 2, 3 | 323 | |||
AW166143.1 | - | 2, 3 | 308 | |||
AW888225.1 | - | 2, 3 | 207 | |||
AY426672.1 | - | 1, 2 | 542 | |||
BC053983.1 | - | 1 | 472 | 5470630 | ||
BC094776.1 | - | 1 | 443 | |||
BE502793.1 | - | 2, 3 | 323 | |||
BQ109532.1 | - | 1, 2 | 83 | |||
CB993617.1 | - | 470 | ||||
CK300836.1 | - | 2, 3 | 182 | |||
DA065905.1 | - | 2, 3 | 289 | |||
DA362466.1 | - | 2, 3 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
