siRNA Details
Gene Symbol: PLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170585.1 | NP_001164056.1 | 16 | 1125 | ||
NM_153021.4 | NP_694566.4 | 16 | 1092 | |||
XM_011532579.2 | XP_011530881.1 | 18 | 1399 | |||
XM_011532581.2 | XP_011530883.1 | 18 | 1362 | |||
XM_011532584.2 | XP_011530886.1 | 18 | 1399 | |||
XM_011532588.2 | XP_011530890.1 | 18 | 1401 | |||
XM_011532589.2 | XP_011530891.1 | 18 | 1396 | |||
XM_011532590.2 | XP_011530892.1 | 18 | 1401 | |||
XM_011532591.2 | XP_011530893.1 | 18 | 1398 | |||
XM_011532593.2 | XP_011530895.1 | 14 | 974 | |||
XM_011532601.2 | XP_011530903.1 | 4 | 387 | |||
XM_011532602.2 | XP_011530904.1 | 3 | 359 | |||
XM_011532603.2 | XP_011530905.1 | 3 | 436 | |||
XM_011532606.2 | XP_011530908.1 | 18 | 1421 | |||
XM_011532607.2 | XP_011530909.1 | 18 | 1421 | |||
XM_011532608.2 | XP_011530910.1 | 18 | 1419 | |||
XM_011532609.2 | XP_011530911.1 | 3 | 357 | |||
XM_017003432.1 | XP_016858921.1 | 17 | 1368 | |||
XM_017003433.1 | XP_016858922.1 | 17 | 1334 | |||
XM_017003434.1 | XP_016858923.1 | 18 | 1403 | |||
GenBank mRNA | BC065041.1 | - | 3 | 178 | ||
BC150633.1 | - | 16 | 1092 | 9057003 | ||
BC153864.1 | - | 16 | 1077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
