siRNA Details
Gene Symbol: PLAUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005377.2 | NP_001005377.1 | 6 | 1087 | ||
NM_001301037.1 | NP_001287966.1 | 6 | 1075 | |||
NM_002659.3 | NP_002650.1 | 7 | 1222 | |||
XM_011527027.2 | XP_011525329.1 | 6 | 900 | |||
XM_011527028.2 | XP_011525330.1 | 7 | 1064 | |||
XM_011527030.2 | XP_011525332.1 | 5 | 762 | |||
XM_011527031.2 | XP_011525333.1 | 6 | 917 | |||
XM_017026872.1 | XP_016882361.1 | 7 | 1457 | |||
XM_017026873.1 | XP_016882362.1 | 5 | 1110 | |||
GenBank mRNA | A18757.1 | - | 7 | 1042 | ||
AK290774.1 | - | 7 | 1042 | |||
BC002788.2 | - | 7 | 1061 | 3617894 | ||
M83246.1 | - | 7 | 1013 | |||
U08839.1 | - | 6 | 1287 | |||
X51675.1 | - | 7 | 1042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
