siRNA Details
Gene Symbol: LY6G6F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003693.1 | NP_001003693.1 | 2 | 244 | ||
GenBank mRNA | AJ496460.1 | - | 2 | 244 | ||
BC137212.1 | - | 2 | 288 | 9021209 | ||
BC137213.1 | - | 2 | 288 | 9021210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
