siRNA Details
Gene Symbol: NFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198989.1 | NP_001185918.1 | 4, 5 | 493 | ||
NM_021100.4 | NP_066923.3 | 5, 6 | 646 | |||
NR_037570.2 | 6, 7 | 768 | ||||
GenBank mRNA | AF097025.2 | - | 5, 6 | 632 | ||
AJ010952.1 | - | 1, 2 | 154 | |||
AK001265.1 | - | 5, 6 | 632 | |||
AK001470.1 | - | 5, 6 | 642 | |||
AK056242.1 | - | 5, 6 | 917 | |||
AK223239.1 | - | 5, 6 | 632 | |||
AK297969.1 | - | 6, 7 | 755 | |||
AK302023.1 | - | 4, 5 | 480 | |||
AK312293.1 | - | 5, 6 | 633 | |||
BC065560.1 | - | 5, 6 | 618 | 6101221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
