siRNA Details
Gene Symbol: MTTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000253.3 | NP_000244.2 | 17 | 2556 | ||
NM_001300785.1 | NP_001287714.1 | 16 | 2394 | |||
GenBank mRNA | AK290793.1 | - | 16 | 2363 | ||
AK302602.1 | - | 16 | 2391 | |||
BC125110.1 | - | 16 | 2335 | 40118807 | ||
BC125111.1 | - | 16 | 2335 | 40118809 | ||
X59657.1 | - | 16 | 2381 | |||
X75500.1 | - | 16 | 2319 | |||
X91148.1 | - | 16 | 2320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
