siRNA Details
Gene Symbol: EDNRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000115.3 | NP_000106.1 | 4 | 987 | ||
NM_001122659.2 | NP_001116131.1 | 3 | 1011 | |||
NM_001201397.1 | NP_001188326.1 | 4 | 1176 | |||
NM_003991.3 | NP_003982.1 | 3 | 1011 | |||
GenBank mRNA | AB209198.1 | - | 3 | 990 | ||
AF114165.1 | - | 4 | 1173 | |||
AK290699.1 | - | 3 | 1011 | |||
AY275463.1 | - | 3 | 753 | |||
BC014472.1 | - | 3 | 988 | 4868863 | ||
D90402.1 | - | 4 | 990 | |||
L06623.1 | - | 3 | 986 | |||
M74921.1 | - | 3 | 799 | |||
S44866.1 | - | 3 | 983 | |||
S57283.1 | - | 4 | 990 | |||
X99250.1 | - | 3 | 986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
