siRNA Details
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001928.2 | NP_001001928.1 | 5, 6 | 697 | ||
NM_005036.4 | NP_005027.2 | 6, 7 | 781 | |||
XM_005261655.3 | XP_005261712.1 | 6, 7 | 1074 | |||
XM_005261656.3 | XP_005261713.1 | 7, 8 | 1238 | |||
XM_006724269.3 | XP_006724332.1 | 6, 7 | 840 | |||
XM_006724270.3 | XP_006724333.1 | 5, 6 | 746 | |||
XM_011530239.2 | XP_011528541.1 | 7, 8 | 930 | |||
XM_011530240.2 | XP_011528542.1 | 7, 8 | 933 | |||
XM_011530241.2 | XP_011528543.1 | 6, 7 | 838 | |||
XM_011530242.2 | XP_011528544.1 | 5, 6 | 660 | |||
XM_011530243.2 | XP_011528545.1 | 4, 5 | 901 | |||
GenBank mRNA | AB307690.1 | - | 3, 4 | 515 | ||
AK289821.1 | - | 6, 7 | 1074 | |||
BC000052.2 | - | 5, 6 | 725 | |||
CR456547.1 | - | 4, 5 | 651 | |||
CR457435.1 | - | 3, 4 | 515 | |||
EU650667.1 | - | 3, 4 | 515 | |||
HQ692862.1 | - | 3, 4 | 515 | |||
L02932.1 | - | 5, 6 | 731 | |||
S74349.1 | - | 4, 5 | 638 | |||
Y07619.1 | - | 3, 4 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
