siRNA Details
Gene Symbol: Marf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081154.2 | NP_001074623.1 | 7 | 1656 | ||
GenBank mRNA | AK035704.1 | - | 1310 | |||
AK220341.1 | - | 4 | 3320 | |||
BC069852.1 | - | 3 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
