siRNA Details
Gene Symbol: Nova1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021361.1 | NP_067336.1 | 3 | 451 | ||
GenBank mRNA | AF232828.1 | - | 3 | 385 | ||
AK090295.1 | - | 3 | 740 | |||
BM942596.1 | - | 3 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
