siRNA Details
Gene Symbol: Nol8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271397.1 | NP_001258326.1 | 16 | 3973 | ||
GenBank mRNA | AK017551.1 | - | 9 | 1047 | ||
AK147172.1 | - | 16 | 3709 | |||
BC026402.1 | - | 3 | 1764 | |||
BC058699.1 | - | 16 | 4129 | |||
BC094941.1 | - | 8 | 843 | |||
BC125575.1 | - | 16 | 3699 | 40130090 | ||
BC125579.1 | - | 16 | 3699 | 40130094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
