siRNA Details
Gene Symbol: Mrpl44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081210.1 | NP_001074679.1 | 2 | 577 | ||
GenBank mRNA | AK007652.1 | - | 1 | 330 | ||
AK019986.1 | - | 2 | 480 | |||
AK146024.1 | - | 2 | 389 | |||
AK150537.1 | - | 2 | 339 | |||
AK151854.1 | - | 2 | 339 | |||
AK156814.1 | - | 2 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
