siRNA Details
Gene Symbol: Mrpl44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081210.1 | NP_001074679.1 | 4 | 1338 | ||
GenBank mRNA | AK007652.1 | - | 3 | 1091 | ||
AK146024.1 | - | 4 | 1150 | |||
AK150537.1 | - | 4 | 1100 | |||
AK151854.1 | - | 4 | 1100 | |||
AK156814.1 | - | 4 | 1253 | |||
BC019727.1 | - | 2 | 571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
