siRNA Details
Gene Symbol: Tdrkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028307.1 | NP_082583.1 | 2 | 98 | ||
GenBank mRNA | BC031886.1 | - | 2 | 92 | ||
BC049363.1 | - | 2 | 98 | 6490411 | ||
BC057030.1 | - | 2 | 114 | 6833743 |

More Information
Other Species Targeted

Gene Ontology Categories:
