siRNA Details
Gene Symbol: Tdrkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028307.1 | NP_082583.1 | 5, 6 | 641 | ||
GenBank mRNA | AK012596.1 | - | 5, 6 | 682 | ||
BC031886.1 | - | 5, 6 | 535 | |||
BC049363.1 | - | 5, 6 | 641 | 6490411 | ||
BC057030.1 | - | 5, 6 | 657 | 6833743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
