siRNA Details
Gene Symbol: Tnrc6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198022.2 | NP_932139.2 | 18, 19 | 5379 | ||
GenBank mRNA | AK036989.1 | - | 14, 15 | 2858 | ||
AK047249.1 | - | 2, 3 | 265 | |||
AK122529.1 | - | 14, 15 | 3330 | |||
AK134064.1 | - | 2, 3 | 256 | |||
AK147471.1 | - | 19, 20 | 5377 | |||
AK147551.1 | - | 17, 18 | 8254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
