siRNA Details
Gene Symbol: Tnrc6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198022.2 | NP_932139.2 | 22 | 8102 | ||
GenBank mRNA | AK137381.1 | - | 1 | 428 | ||
AK147551.1 | - | 21 | 10975 | |||
AK166722.1 | - | 1 | 1359 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
