siRNA Details
Gene Symbol: Tnrc6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198022.2 | NP_932139.2 | 12, 13 | 4430 | ||
GenBank mRNA | AK036989.1 | - | 8, 9 | 1909 | ||
AK122529.1 | - | 8, 9 | 2381 | |||
AK147471.1 | - | 13, 14 | 4428 | |||
AK147551.1 | - | 12, 13 | 4146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
