siRNA Details
Gene Symbol: Rbm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045807.1 | NP_001039272.1 | 1 | 1816 | ||
GenBank mRNA | AK048987.1 | - | 1051 | |||
AK077048.1 | - | 1 | 2204 | |||
AK166271.1 | - | 1 | 1579 | |||
AK168242.1 | - | 1 | 1577 | |||
AK168936.1 | - | 1 | 1701 | |||
AK220162.1 | - | 1 | 1628 | |||
BC051409.1 | - | 1 | 1570 | |||
BC057038.1 | - | 1 | 1580 | |||
BC080828.1 | - | 1 | 1561 | |||
BC120590.1 | - | 1 | 1816 | 40129513 | ||
BC137741.1 | - | 1 | 1816 | 8860760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
