siRNA Details
Gene Symbol: Rbm27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172626.2 | NP_766214.2 | 11 | 1851 | ||
GenBank mRNA | AK042613.1 | - | 7 | 1249 | ||
AK129330.1 | - | 6 | 860 | |||
AY461716.1 | - | 11 | 1826 | |||
BC054080.1 | - | 8 | 1464 | 6390990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
