siRNA Details
Gene Symbol: Rbm27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172626.2 | NP_766214.2 | 20 | 4196 | ||
GenBank mRNA | AK033739.1 | - | 3 | 1290 | ||
AK041494.1 | - | 1 | 407 | |||
AK042456.1 | - | 1 | 408 | |||
AK087759.1 | - | 7 | 1778 | |||
AK129330.1 | - | 15 | 3205 | |||
BC042631.1 | - | 3 | 1379 | |||
BC054080.1 | - | 16 | 3512 | 6390990 | ||
BC059854.1 | - | 5 | 1614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
