siRNA Details
Gene Symbol: Snrpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027246.1 | NP_081522.1 | 4 | 485 | ||
GenBank mRNA | AK135835.1 | - | 4 | 481 | ||
BC066015.1 | - | 4 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
