siRNA Details
Gene Symbol: Rbm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025776.2 | NP_080052.1 | 7 | 648 | ||
GenBank mRNA | AK018452.1 | - | 7 | 671 | ||
AK045116.1 | - | 7 | 668 | |||
AK150271.1 | - | 7 | 661 | |||
AK151758.1 | - | 7 | 661 | |||
AK152252.1 | - | 7 | 661 | |||
AK165277.1 | - | 7 | 666 | |||
AK166600.1 | - | 7 | 669 | |||
AK167509.1 | - | 7 | 688 | |||
AK168919.1 | - | 7 | 649 | |||
BC042691.1 | - | 7 | 667 | |||
BC080205.1 | - | 7 | 646 | 3709760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
