siRNA Details
Gene Symbol: Tnrc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144925.3 | NP_659174.3 | 24 | 6158 | ||
GenBank mRNA | AK044679.1 | - | 12 | 2542 | ||
AK050601.1 | - | 6 | 1038 | |||
AK138426.1 | - | 14 | 2208 | |||
AK142808.1 | - | 9 | 1527 | |||
AK142809.1 | - | 9 | 1527 | |||
AK147327.1 | - | 24 | 5863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
