siRNA Details
Gene Symbol: Tnrc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144925.3 | NP_659174.3 | 24 | 8244 | ||
GenBank mRNA | AK014218.1 | - | 1 | 699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
