siRNA Details
Gene Symbol: Adad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029428.1 | NP_083704.1 | 6 | 984 | ||
GenBank mRNA | AK015063.1 | - | 6 | 984 | ||
BC137980.1 | - | 6 | 971 | 8860999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
