siRNA Details
Gene Symbol: Dus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301176.1 | NP_001288105.1 | 10 | 747 | ||
NM_025518.4 | NP_079794.1 | 11 | 791 | |||
GenBank mRNA | AK009391.1 | - | 11 | 763 | ||
AK077451.1 | - | 10 | 719 | |||
BC058431.1 | - | 2 | 62 | |||
BC116934.1 | - | 11 | 709 | 40126253 | ||
BC116936.1 | - | 11 | 709 | 40126255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
