siRNA Details
Gene Symbol: A1cf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081074.1 | NP_001074543.1 | 4 | 441 | ||
GenBank mRNA | AK007970.1 | - | 3 | 285 | ||
AY566862.1 | - | 4 | 438 | |||
AY566863.1 | - | 4 | 438 | |||
AY566864.1 | - | 4 | 438 | |||
BC038942.1 | - | 2 | 141 | |||
BC089622.1 | - | 4 | 389 | 6760455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
