siRNA Details
Gene Symbol: RAB40C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172663.1 | NP_001166134.1 | 7 | 2562 | ||
NM_001172664.1 | NP_001166135.1 | 7 | 2454 | |||
NM_001172665.1 | NP_001166136.1 | 7 | 2414 | |||
NM_001172666.1 | NP_001166137.1 | 7 | 2357 | |||
NM_021168.4 | NP_066991.3 | 6 | 2459 | |||
GenBank mRNA | AK000584.1 | - | 1 | 794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
