siRNA Details
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021185.4 | NP_067008.3 | 19 | 2381 | ||
XM_005259114.4 | XP_005259171.1 | 19 | 2477 | |||
XM_005259115.4 | XP_005259172.1 | 19 | 2400 | |||
XM_005259116.2 | XP_005259173.1 | 17 | 2201 | |||
XM_006723308.3 | XP_006723371.1 | 19 | 2997 | |||
XM_006723310.2 | XP_006723373.1 | 19 | 2264 | |||
XM_006723311.2 | XP_006723374.1 | 19 | 2402 | |||
XM_011527174.2 | XP_011525476.1 | 19 | 2391 | |||
XM_011527175.2 | XP_011525477.1 | 19 | 2297 | |||
XM_011527176.2 | XP_011525478.1 | 19 | 2300 | |||
XM_011527177.2 | XP_011525479.1 | 19 | 2261 | |||
XM_011527178.2 | XP_011525480.1 | 20 | 2743 | |||
XM_011527179.1 | XP_011525481.1 | 19 | 2381 | |||
XM_011527182.2 | XP_011525484.1 | 18 | 2125 | |||
XM_017027053.1 | XP_016882542.1 | 18 | 2261 | |||
XM_017027054.1 | XP_016882543.1 | 17 | 1994 | |||
XM_017027055.1 | XP_016882544.1 | 6 | 1470 | |||
GenBank mRNA | AK128220.1 | - | 19 | 2164 | ||
AK310460.1 | - | 15 | 1711 | |||
BC094785.1 | - | 19 | 2301 | |||
BC109035.1 | - | 15 | 1780 | 40028284 |
