siRNA Details
Gene Symbol: KIR2DL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020535.3 | NP_065396.1 | 4 | 575 | ||
GenBank mRNA | AF204903.1 | - | 4 | 575 | ||
AF217487.1 | - | 6 | 544 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
