siRNA Details
Gene Symbol: BAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182481.1 | NP_872287.1 | 4 | 519 | ||
GenBank mRNA | AF339514.1 | - | 4 | 519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
