siRNA Details
Gene Symbol: IGLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020070.3 | NP_064455.1 | 1 | 229 | ||
NM_152855.2 | NP_690594.1 | 1 | 229 | |||
XM_011530169.2 | XP_011528471.1 | 1 | 469 | |||
GenBank mRNA | BC012293.1 | - | 1 | 169 | 4508892 | |
BC030239.1 | - | 1 | 171 | |||
M27749.1 | - | 1 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
