siRNA Details
Gene Symbol: CLEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002975.2 | NP_002966.1 | 4 | 1321 | ||
GenBank mRNA | AF020044.1 | - | 4 | 1322 | ||
BC005810.2 | - | 4 | 1285 | 3830506 | ||
D86586.1 | - | 5 | 1048 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
