siRNA Details
Gene Symbol: MZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267033.1 | NP_001253962.1 | 6 | 2663 | ||
NM_003422.2 | NP_003413.2 | 6 | 2589 | |||
NM_198055.1 | NP_932172.1 | 6 | 2907 | |||
XM_005259204.3 | XP_005259261.1 | 7 | 3262 | |||
XM_017027206.1 | XP_016882695.1 | 6 | 3110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
