siRNA Details
Gene Symbol: LMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142315.1 | NP_001135787.1 | 4 | 1291 | ||
NM_001142316.1 | NP_001135788.1 | 3 | 1391 | |||
NM_005574.3 | NP_005565.2 | 6 | 1989 | |||
XM_005252921.2 | XP_005252978.1 | 5 | 1874 | |||
XM_017017727.1 | XP_016873216.1 | 5 | 1642 | |||
XM_017017728.1 | XP_016873217.1 | 7 | 1743 | |||
XM_017017729.1 | XP_016873218.1 | 6 | 1626 | |||
XM_017017730.1 | XP_016873219.1 | 5 | 1808 | |||
XM_017017731.1 | XP_016873220.1 | 6 | 1589 | |||
XM_017017732.1 | XP_016873221.1 | 5 | 1520 | |||
XM_017017733.1 | XP_016873222.1 | 5 | 1427 | |||
GenBank mRNA | AF257211.1 | - | 2 | 1245 | ||
AK127915.1 | - | 1 | 1676 | |||
BC034041.1 | - | 3 | 1393 | 4796599 | ||
BC035607.1 | - | 3 | 1279 | 5186480 | ||
BC042426.1 | - | 3 | 1382 | 5188482 | ||
BC073973.1 | - | 3 | 1214 | 5734404 | ||
X61118.1 | - | 6 | 1988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
