siRNA Details
Gene Symbol: ZFP36L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006887.4 | NP_008818.3 | 2 | 1432 | ||
GenBank mRNA | BC005010.1 | - | 2 | 1221 | 3640393 | |
X78992.1 | - | 2 | 1201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
