siRNA Details
Gene Symbol: NFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198989.1 | NP_001185918.1 | 12 | 1761 | ||
NM_021100.4 | NP_066923.3 | 13 | 1914 | |||
NR_037570.2 | 14 | 2036 | ||||
GenBank mRNA | AF097025.2 | - | 13 | 1899 | ||
AJ010952.1 | - | 9 | 1422 | |||
AK001265.1 | - | 13 | 1899 | |||
AK001470.1 | - | 13 | 1910 | |||
AK056242.1 | - | 13 | 2185 | |||
AK091049.1 | - | 5 | 2456 | |||
AK223239.1 | - | 13 | 1900 | |||
BC065560.1 | - | 13 | 1886 | 6101221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
