siRNA Details
Gene Symbol: RTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021136.2 | NP_066959.1 | 2 | 970 | ||
XM_011537063.2 | XP_011535365.1 | 2 | 1419 | |||
GenBank mRNA | BC090862.1 | - | 2 | 885 | 6179803 | |
BC111694.1 | - | 3 | 660 | 40035553 | ||
L10333.1 | - | 2 | 883 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
