siRNA Details
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206878.1 | NP_001193807.1 | 7 | 1726 | ||
NM_021198.2 | NP_067021.1 | 7 | 1990 | |||
NM_182642.2 | NP_872580.1 | 7 | 1987 | |||
XM_011511569.2 | XP_011509871.1 | 7 | 2137 | |||
XM_011511570.2 | XP_011509872.1 | 7 | 2133 | |||
GenBank mRNA | AF217969.1 | - | 5 | 1394 | ||
AF229162.1 | - | 7 | 1990 | |||
AF258582.1 | - | 5 | 1450 | |||
AK075090.1 | - | 2 | 1116 | |||
BC012977.2 | - | 7 | 1699 | 3687816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
