siRNA Details
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136219.1 | NP_001129691.1 | 6, 7 | 826 | ||
NM_021642.3 | NP_067674.2 | 6, 7 | 823 | |||
XM_011509287.2 | XP_011507589.1 | 6, 7 | 811 | |||
XM_011509290.2 | XP_011507592.1 | 6, 7 | 811 | |||
XM_017000663.1 | XP_016856152.1 | 6, 7 | 808 | |||
XM_017000664.1 | XP_016856153.1 | 6, 7 | 811 | |||
XM_017000665.1 | XP_016856154.1 | 6, 7 | 811 | |||
XM_017000666.1 | XP_016856155.1 | 5, 6 | 688 | |||
XM_017000667.1 | XP_016856156.1 | 5, 6 | 688 | |||
XM_017000668.1 | XP_016856157.1 | 5, 6 | 685 | |||
GenBank mRNA | AK225438.1 | - | 6, 7 | 784 | ||
AK225601.1 | - | 6, 7 | 779 | |||
AK226059.1 | - | 6, 7 | 795 | |||
AK298657.1 | - | 6, 7 | 897 | |||
AK298734.1 | - | 5, 6 | 384 | |||
AK315225.1 | - | 6, 7 | 784 | |||
BC019931.1 | - | 6, 7 | 808 | 4536764 | ||
BC020823.1 | - | 6, 7 | 773 | 4704252 | ||
J03619.1 | - | 6, 7 | 778 | |||
M28697.1 | - | 6, 7 | 760 | |||
M31932.1 | - | 6, 7 | 780 | |||
X62572.1 | - | 6, 7 | 760 | |||
Y00644.1 | - | 6, 7 | 792 |
Gene Symbol: FCGR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201563.5 | NP_963857.3 | 6, 7 | 890 | ||
NR_047648.1 | 6, 7 | 890 | ||||
GenBank mRNA | BC137397.1 | - | 6, 7 | 833 | 9021395 | |
BC137423.1 | - | 6, 7 | 833 | 9021421 | ||
U90938.1 | - | 6, 7 | 793 | |||
X17652.1 | - | 6, 7 | 824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
