siRNA Details
Gene Symbol: UTP14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021645.5 | NP_067677.4 | 2 | 1080 | ||
GenBank mRNA | BC089407.1 | - | 1 | 489 | 3065365 | |
BC122536.1 | - | 2 | 857 | 40036054 | ||
D87455.1 | - | 2 | 1080 |
Gene Symbol: ALG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004127.2 | NP_001004127.2 | 4 | 2072 | ||
NR_036571.2 | 2 | 909 | ||||
GenBank mRNA | AK025456.1 | - | 4 | 2045 | ||
BC073862.1 | - | 2 | 1228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
