siRNA Details
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258001.1 | NP_001244930.1 | 5 | 423 | ||
NM_001258003.1 | NP_001244932.1 | 4 | 330 | |||
NM_001258004.1 | NP_001244933.1 | 5 | 423 | |||
NM_021797.3 | NP_068569.2 | 3 | 300 | |||
NM_201653.3 | NP_970615.2 | 6 | 625 | |||
XM_017001048.1 | XP_016856537.1 | 4 | 333 | |||
GenBank mRNA | AB025008.1 | - | 3 | 290 | ||
AF290004.1 | - | 6 | 561 | |||
AK098814.1 | - | 3 | 936 | |||
AK290702.1 | - | 5 | 425 | |||
AY789444.1 | - | 5 | 359 | |||
AY825504.1 | - | 4 | 320 | |||
AY911310.1 | - | 4 | 279 | |||
BC036339.1 | - | 3 | 238 | 5184463 | ||
BC047336.1 | - | 4 | 247 | 5185486 | ||
BC106910.1 | - | 2 | 206 | 40031872 | ||
BC139901.1 | - | 4 | 252 | |||
BC139920.1 | - | 4 | 252 |
Gene Symbol: CHIAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003928.2 | 3 | 1099 | |||
GenBank mRNA | BC031662.2 | - | 3 | 1099 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023186, AK008633, AK008650, BC011134, AK007573, and others.
Gene Ontology Categories:
