siRNA Details
Gene Symbol: ARHGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK160365.1 | - | 10 | 4280 | ||
BC010046.2 | - | 10 | 1169 | |||
BC011839.2 | - | 10 | 1169 | |||
BC014555.2 | - | 10 | 1169 | |||
BC136661.1 | - | 10 | 4138 | 9020651 | ||
BX649178.1 | - | 9 | 2109 | |||
CR749203.1 | - | 10 | 4164 | |||
U02082.1 | - | 8 | 965 |
Gene Symbol: ARHGEF34P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033942.1 | 9 | 3142 | |||
GenBank mRNA | BC107871.1 | - | 3 | 163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
