siRNA Details
Gene Symbol: BPESC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026783.3 | 1 | 589 | |||
GenBank mRNA | AF196865.1 | - | 1 | 589 | ||
BC119686.1 | - | 1 | 573 | |||
BC119687.1 | - | 1 | 573 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BPESC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026783.3 | 1 | 589 | |||
GenBank mRNA | AF196865.1 | - | 1 | 589 | ||
BC119686.1 | - | 1 | 573 | |||
BC119687.1 | - | 1 | 573 |